Learn More
Saint-Gobain Plastic Sanitary Couplings
Click to view available options
Maximum Pressure:
200 psi
Unit Size:
Each
Description
- Reduces dead space and improves flow.
- Tapered funnel design provides a steady fluid flow.
- Smooth inner surface prevents particle entrapment.
- Seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth.
- Material meets USP Class VI requirements and is FDA compliant for food contact.
Specifications
Specifications
| Certifications/Compliance | USP Class VI |
| Autoclavable | Not Autoclavable |
| Material | Polypropylene |
| For Use With (Application) | For bioreactors and fermentors |
| Min. Operating Temperature (English) | 72°F |
| Min. Operating Temperature (Metric) | 22°C |
| Hose Barb | Fitting Tygopure 3/8 in. Polypropylene |
| Maximum Pressure | 200 psi |
| Max. Operating Temperature (English) | 200°F |
| Max. Operating Temperature (Metric) | 93°C |
| Show More |