Learn More
Microspec™ Plastic Inoculation Loops
Product Code. 16634822
Click to view available options
Volume:
1 μL
5 μL
10 μL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 250
Product Code. 16634822
Brand: Microspec™ B5
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Available as aseptic manufactured or ethylene oxide sterilized.
  • Includes 250 Plastic Inoculation Loops
  • Aseptic Manufactured or Ethylene Oxide Sterilized
  • 200 mm length
Specifications
Disposable Single-use
Volume 5 μL
Sterility Non-sterile
Color Blue
Length (Metric) Handle 200 mm
Quantity 250
Packaging Type Microseal Pouch
Calibrated Yes