Learn More
Fisherbrand™ Plastic Disposable Generator Probe
Shop All Fisherbrand Products
Click to view available options
Packaging Quantity:
100 Pk.
25 Pk.
50 Pk.
Unit Size:
Pack of 100
Pack of 25
Pack of 50
Description
- Ideal for tough to process samples
- Economical: <1% of cost of stainless steel generator probes
- Eliminates cross contamination
- Ideal for PCR, RNA and DNA extractions
- Maintenance free
- Processing range: >250 μL - maximum volume will be application specific
Specifications
Specifications
| Product Type | Probe |
| Diameter (English) Probe | 12 mm |
| Length (English) Probe | 110 mm |
| Autoclavable | Autoclavable |
| Blade Material | Polypropylene |
| Material | Polycarbonate, Polyetherimide |
| Closure Color | Blue |
| For Use With (Equipment) | Fisherbrand 150 and 850 Homogenizer |
| Packaging Quantity | 50 Pk. |