Learn More
Plasmid DNA isolation solution I, Thermo Scientific™
Product Code. 15443879
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Product Code. 15443879
Brand: Thermo Scientific Alfa Aesar J61263.AP
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Plasmid DNA isolation solution I
Quantity 500 mL
Physical Form Liquid