Learn More
Invitrogen™ PLA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139916
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PVHSTMPGTV TSSCSPGNNS RPSSPGSGLH ASSPTASQNN SKAAVNSASS FNSSGSWYRW NHSTYL Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 90%.
This gene encodes a member of the POU domain family of transcription factors. POU domain transcription factors bind to a specific octamer DNA motif and regulate cell type-specific differentiation pathways. The encoded protein is primarily expressed in the epidermis, and plays a critical role in keratinocyte proliferation and differentiation. The encoded protein is also a candidate tumor suppressor protein, and aberrant promoter methylation of this gene may play a role in cervical cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Sep 2011].
Specifications
| PLA1 | |
| Polyclonal | |
| Unconjugated | |
| POU2F3 | |
| 11-Oct; Epoc-1; FLJ40063; LOC553427 protein; MGC126698; Oct11; oct-11; Oct-11a; octamer-binding protein 11; Octamer-binding transcription factor 11; OTF11; OTF-11; PLA1; PLA-1; POU class 2 homeobox 3; POU domain transcription factor OCT11a; POU domain, class 2, transcription factor 3; POU transcription factor; POU2F3; Rov 1 pou-homeodomain protein; Skin; Skin-1a; Skn-1a; Skn-li; Transcription factor PLA-1; transcription factor Skn-1; zgc:136377; zgc:158534 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25833 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UKI9 | |
| POU2F3 | |
| Recombinant protein corresponding to Human POU2F3. Recombinant protein control fragment (Product #RP-109346). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.