missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC mu [p Ser916] Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38645-20ul
This item is not returnable.
View return policy
Description
PKC mu Polyclonal antibody specifically detects PKC mu in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| PKC mu | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 2.7.11, nPKC-D1, nPKC-mu, PKCM, PKC-MU, PKD1, PKDEC 2.7.11.13, PRKCMPKC-mu, Protein kinase C mu type, protein kinase C, mu, Protein kinase D, protein kinase D1, serine/threonine-protein kinase D1 | |
| A synthetic phosphorylated peptide around S912 of human PKC mu (NP_002733.2).,, Sequence:, RVSIL | |
| 20 μL | |
| Golgi Apparatus Markers, Phospho Specific, Signal Transduction | |
| 5587 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction