missing translation for 'onlineSavingsMsg'
Learn More

PKC gamma Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2F4B9, Proteintech

Mouse Monoclonal Antibody

145.00 EUR - 410.00 EUR

Specifications

Antigen PKC gamma
Clone 2F4B9
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16898295
View Documents
Proteintech
66429-1-IG-20UL
20 μL
145.00 EUR
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16888295
View Documents
Proteintech
66429-1-IG-150UL
150 μL
410.00 EUR
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.
Especificaciones

Especificaciones

PKC gamma
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
PKC gamma, PKCC, PKCG, PRKCG, Protein kinase C gamma type, protein kinase C, gamma, SCA14
Prkcg
IgG2a
Protein A
Antibody
2F4B9
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Mouse
Pig, Mouse, Human, Rat
P05129, P63318, P63319
100518074, 18752, 24681, 5582
PKC gamma Fusion Protein Ag5910
Primary
-20°C
PRKCG
Videos
SDS
Documentos

Documentos

Product Certifications
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado