missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC epsilon Mouse anti-Human, Clone: 666843, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB51341-SP
This item is not returnable.
View return policy
Description
PKC epsilon Monoclonal specifically detects PKC epsilon in Human samples. It is validated for Western Blot.Specifications
| PKC epsilon | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EC 2.7.11, EC 2.7.11.13, MGC125656, nPKC-epsilon, PKCE, PKCEMGC125657, PRKCE, protein kinase C epsilon type, protein kinase C, epsilon | |
| E. coli-derived recombinant human PKC epsilon Gln580-Pro737 Accession # Q02156 | |
| 25 μg | |
| Primary | |
| Detects human PKC epsilon in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 666843 | |
| Western Blot 2 ug/mL | |
| Q02156 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5581 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
RUO