missing translation for 'onlineSavingsMsg'
Learn More

PK11000, MedChemExpress

Product Code. p-200097159
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16434116

Brand: Medchem Express HYU00447

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PK11000 is an alkylating agent, and stabilizes the DNA-binding domain of both WT and mutant p53 proteins by covalent cysteine modification without compromising DNA binding. PK11000 has anti-tumor activities.

PK11000 is an alkylating agent, and stabilizes the DNA-binding domain of both WT and mutant p53 proteins by covalent cysteine modification without compromising DNA binding. PK11000 has anti-tumor activities.

Specifications

Chemical Name or Material PK11000
CAS 38275-34-2
Color White
Purity Grade Notes Research
Molecular Formula C6H5ClN2O4S
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (422.60 mM; Need ultrasonic) ∣H2O : 25 mg/mL (105.65 mM; Need ultrasonic)
SMILES O=C(C1=NC(S(=O)(C)=O)=NC=C1Cl)O
Molecular Weight (g/mol) 236.63
Formula Weight 236.63
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.