Learn More
PK11000, MedChemExpress
Product Code. 16434116
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. 16434116
Brand: Medchem Express HYU00447
MedChemExpress PK11000 is an alkylating agent, and stabilizes the DNA-binding domain of both WT and mutant p53 proteins by covalent cysteine modification without compromising DNA binding. PK11000 has anti-tumor activities.
PK11000 is an alkylating agent, and stabilizes the DNA-binding domain of both WT and mutant p53 proteins by covalent cysteine modification without compromising DNA binding. PK11000 has anti-tumor activities.
Specifications
Chemical Name or Material PK11000
CAS 38275-34-2
Color White
Purity Grade Notes Research
Molecular Formula C6H5ClN2O4S
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (422.60 mM; Need ultrasonic) ∣H2O : 25 mg/mL (105.65 mM; Need ultrasonic)
SMILES O=C(C1=NC(S(=O)(C)=O)=NC=C1Cl)O
Molecular Weight (g/mol) 236.63
Formula Weight 236.63
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death