missing translation for 'onlineSavingsMsg'
Learn More

Pirozadil, MedChemExpress

Product Code. 16495537
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495537 5 mg 5mg
16482526 25 mg 25mg
16462798 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16495537 Supplier Medchem Express Supplier No. HY100144

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pirozadil is a hypolipidemic agent.

Pirozadil is a hypolipidemic agent.

Specifications

Chemical Name or Material Pirozadil
CAS 54110-25-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H29NO10
Quantity 5 mg
Solubility Information DMSO : 62.5 mg/mL (118.48 mM; Need ultrasonic)
SMILES O=C(OCC1=CC=CC(COC(C2=CC(OC)=C(OC)C(OC)=C2)=O)=N1)C3=CC(OC)=C(OC)C(OC)=C3
Molecular Weight (g/mol) 527.52
Formula Weight 527.52
Percent Purity 98.31%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.