Learn More
Eppendorf™ Pipette Holder 2 for one Multipette™ E3/E3x or Multipette™ stream/Xstream Pipette
Shop All Eppendorf Products
Description
- Carries one Multipette™ E3/E3x or Multipette™ stream/Xstream
- Same as Charger Shell 2 for Multipette E3/E3x or Multipette stream/Xstream but without charging functionality
- Designed for wall mounting or for installation in Pipette Carousel 2
- Sticky tape for wall mounting included
Specifications
Specifications
| For Use With (Equipment) | Multipette™ E3/E3x or Multipette™ stream/Xstream Pipette |