Learn More
Fisherbrand™ Pipette Cone for Accumax Pipette Controller
Product Code. 15850043 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15850043 Green Each
15840043 Blue Each
15860043 Orange Each
15870043 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15850043
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Available in a range of colors.
  • Autoclavable
  • For use with the Accumax Pipette Controller
  • Pick between blue, red, orange or green
Specifications
Color Green
Type Cone
For Use With (Application) Pipetting
For Use With (Equipment) Accumax Pipette Controller