Learn More
Gilson™ PIPETMAN™ G Large-Volume Kit
Shop All Gilson Products
Click to view available options
Volume (Metric):
500 to 10 mL
Unit Size:
1 set
Description
- Low pipetting forces
- Improves comfort and minimizes injury
- Durable
- Accurate
- Features reliability of the original PIPETMAN™ pipettes
Specifications
Specifications
| Material | Polypropylene |
| Product Line | Pipetman G |
| Type | Large Volume Kit |
| Volume (Metric) | 500 to 10 mL |
| Length (Metric) | 33.5 cm |
| Width (Metric) | 22 cm |
| Height (Metric) | 5 cm |
| Weight (Metric) | 0.6 kg |