Learn More
Gilson™ PIPETMANTM Comfort Pipette Handle Stickers
Shop All Gilson Products
Click to view available options
:
Comfort Handle Sticker
Description
- Quick and easy identification of pipettes
- Glossy, pre-cut adhesive labels
Specifications
Specifications
| Content And Storage | 10 sheets of 12 Comfort Handle stickers |
| Color | Dark Blue |
| For Use With | Pipettes |
| Material | Polypropylene |
| Type | Sticker |
| For Use With (Equipment) | PIPETMAN Comfort Pipette Handle |
| Includes | 10 sheets of 12 Comfort Handle stickers |
| Quantity | 120 Labels |
| Dimensions (L x W x H) | 210 X 148.5 mm |
| Length (English) | 8.3 in |
| Show More |