missing translation for 'onlineSavingsMsg'
Learn More

Piperaquine tetraphosphate tetrahydrate, MedChemExpress

Product Code. p-200096331
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16431138 5 mg 5mg
16428728 10 mg 10mg
16424637 50 mg 50mg
16403816 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16431138 Supplier Medchem Express Supplier No. HYB1896B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Piperaquine tetraphosphate tetrahydrate is a bisquinoline antimalarial agent. Piperaquine tetraphosphate tetrahydrate can be used in antimalarial research in combination with Artemisinin.

Piperaquine tetraphosphate tetrahydrate is a bisquinoline antimalarial agent. Piperaquine tetraphosphate tetrahydrate can be used in antimalarial research in combination with Artemisinin.

Specifications

Chemical Name or Material Piperaquine tetraphosphate tetrahydrate
Physical Form Solid
CAS 915967-82-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C29H32Cl2N6.4H3O4P.4H2O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information H2O : 6 mg/mL (6.00 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (5.00 mM; Need ultrasonic and warming)
SMILES ClC1=CC=C2C(N3CCN(CCCN4CCN(C5=CC=NC6=CC(Cl)=CC=C56)CC4)CC3)=CC=NC2=C1.OP(O)(O)=O.[4].[4 H2O]
Molecular Weight (g/mol) 999.6
Formula Weight 999.6
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.