missing translation for 'onlineSavingsMsg'
Learn More

Pinoxaden, TRC

Product Code. 30426459
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426459 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426459 Supplier Toronto Research Chemicals Supplier No. TRCP469505500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pinoxaden
CAS 243973-20-8
Purity Grade Notes HPLC
Molecular Formula C23 H32 N2 O4
Quantity 500 mg
Synonym NewAxial, Pinoxaden
InChI Formula InChI=1S/C23H32N2O4/c1-7-16-13-15(3)14-17(8-2)18(16)19-20(26)24-9-11-28-12-10-25(24)21(19)29-22(27)23(4,5)6/h13-14H,7-12H2,1-6H3
SMILES CCc1cc(C)cc(CC)c1C2=C(OC(=O)C(C)(C)C)N3CCOCCN3C2=O
IUPAC Name [8-(2,6-diethyl-4-methylphenyl)-9-oxo-1,2,4,5-tetrahydropyrazolo[1,2-d][1,4,5]oxadiazepin-7-yl] 2,2-dimethylpropanoate
Molecular Weight (g/mol) 400.51
Formula Weight 400.2362
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.