missing translation for 'onlineSavingsMsg'
Learn More

PIM-447 dihydrochloride, MedChemExpress

Product Code. p-200102149
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493697

Brand: Medchem Express HY19322B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PIM447 dihydrochloride (LGH447 dihydrochloride) is a potent, orally available, and selective pan-PIM kinase inhibitor, with Ki values of 6, 18, and 9 pM for PIM1, PIM2, and PIM3, respectively. PIM447 dihydrochloride displays dual antimyeloma and bone-protective effects. PIM447 dihydrochloride induces apoptosis.

PIM447 dihydrochloride (LGH447 dihydrochloride) is a potent, orally available, and selective pan-PIM kinase inhibitor, with Ki values of 6, 18, and 9 pM for PIM1, PIM2, and PIM3, respectively. PIM447 dihydrochloride displays dual antimyeloma and bone-protective effects. PIM447 dihydrochloride induces apoptosis.

Specifications

Chemical Name or Material PIM-447 dihydrochloride
CAS 1820565-69-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H25Cl2F3N4O
Quantity 50 mg
Synonym LGH447 dihydrochloride
Solubility Information H2O : 50 mg/mL (97.39 mM; Need ultrasonic) ∣DMSO : ≥ 46.7 mg/mL (90.97 mM)
SMILES O=C(C1=NC(C2=C(F)C=CC=C2F)=C(F)C=C1)NC3=C([C@H]4C[C@@H](N)C[C@@H](C)C4)C=CN=C3.Cl.Cl
Molecular Weight (g/mol) 513.38
Formula Weight 513.38
Percent Purity 98.25%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.