missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Piko PCR Plate, 24-well, white

Product Code. p-7184429
Click to view available options
Quantity:
200 plates
50 plates
Unit Size:
Pack of 200
Pack of 50
This item is not returnable. View return policy

Product Code. 10719990

Brand: Thermo Scientific™ SPL0241

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Piko PCR Plates are modular, ultra-thin wall PCR plates for Piko* Thermal Cyclers.

Thermo Scientific™ Piko™ PCR Plates are modular, ultra-thin wall PCR plates for Piko Thermal Cyclers.

Piko PCR Plates are also flexible alternatives for conventional microplates. Four plates, each the size of a microscope slide, can be reversibly snapped into a rigid frame, producing the equivalent of a standard 96- or 384-well microplate. This format allows multiple sample sources to be combined into a single PCR run, or subjected to different post-PCR processing.
  • Wall thickness half that of conventional thin-walled tubes
  • Flexible format saves cost and reduces waste
  • Compatible with standard multi-channel pipettes and liquid handlers
  • Well spacing and footprint correspond to industry (ANSI) dimensions
  • Available in clear and white

Recommended for:

PCR, Fast PCR, High Performance PCR

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified RNase/DNase-free
Product Line Piko
Color White
Product Type PCR Microplate
For Use With (Equipment) Piko Thermal Cycler, PikoReal Real-Time PCR System
Material Polypropylene
No. of Wells 24
Quantity 200 plates
Reaction Speed Low profile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.