missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK-90, MedChemExpress
Description
PIK-90 is a DNA-PK and PI3K inhibitor, which inhibits p110α, p110γ and DNA-PK with IC50s of 11, 18 and 13 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | PIK-90 |
| CAS | 677338-12-4 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H17N5O3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 6.67 mg/mL (18.98 mM; ultrasonic and adjust pH to 2 with HCl) |
| SMILES | O=C(NC1=NC2=C(C3=NCCN13)C=CC(OC)=C2OC)C4=CC=CN=C4 |
| Molecular Weight (g/mol) | 351.36 |
| Formula Weight | 351.36 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?