Learn More
Thermo Scientific™ BCIP Substrate Powder (5-bromo-4-chloro-3'-indolylphosphate p-toluidine)
Shop All Thermo Scientific ProductsDescrizione
Thermo Scientific™ Pierce NBT and BCIP Substrates are powders and ready-made solutions of nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolylphosphate for chromogenic blot and IHC staining with alkaline phosphatase probes.
Highlights:
AP substrate – for detection of alkaline phosphatase (alk-phos) activity on solid media, including nitrocellulose and PVDF membranes and fixed tissue samples
Chromogenic – no special equipment needed for visualization; produces purple-black precipitate that is easy to photograph
Package options – choose individual NBT and BCIP powders, or preformulated, ready-to-use solutions with or without levamisole suppressor
1-Step™ NBT/BCIP
- Ideal for immunoblotting applications
- Minimal assay-to-assay variability
1-Step NBT/BCIP Plus Suppressor
- Ideal for staining tissues assayed with calf intestinal alkaline phosphatase without the interference caused by endogenous phosphatases
- Contains 1mM levamisole to inhibit endogenous phosphatase activity
Specifica
Specifica
| Description | Pierce BCIP Substrate |
| Product Type | Substrate |
| Detection Method | Colorimetric |
| Form | Powder |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Product Line | Pierce™ |
| Quantity | 1 g |
| Substrate | Alkaline Phosphatase |
| Content And Storage | Upon receipt store product at -20°C protected from moisture. Product is shipped at ambient temperature. |
For Research Use Only. Not for use in diagnostic procedures.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.