Learn More
Thermo Scientific™ Pierce™ Goat Anti-Mouse IgG Antibody Coated Plates, Clear, 96-Well
Product Code. 10713944 Shop All Thermo Scientific Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 5
Product Code. 10713944
Brand: Thermo Scientific™ 15134
Perform binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates.
Thermo Scientific™ Pierce Antibody Coated Plates are ideal for binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates.
These plates are pre-coated with goat anti-mouse (GAM) or goat anti-rabbit (GAR) IgG polyclonal antibodies, allowing assay-specific antibodies to be captured even from impure protein samples. The plates are supplied pre-blocked and ready-to-use. Pierce Secondary Antibody Coated Plates can be used for direct, indirect, competitive or sandwich assays and are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.
Highlights:
  • Prevents antibody denaturation caused by direct binding
  • Unlike Protein A or G, these bind only to target species
  • Antibody-binding capacity is higher than direct adsorption
  • Pre-blocked to reduce nonspecific binding
  • Less expensive than Protein A or G plates
TRUSTED_SUSTAINABILITY
Specifications
Binding Property Standard Capacity, Affinity
Content And Storage Note: Plates are activated to 100 μL and supplied pre-blocked with SuperBlock™ Blocking Buffer.
Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag and store desiccated at 4°C.
Detection Method Colorimetric
Packaging 5 Plate
Product Line Pierce
Color Clear
Product Type Detection Plate, Immunoassay, ELISA
Material Polystyrene
Volume (Metric) Well 100 μL
No. of Wells 96
Surface Treatment Antibody
Plate Blocking SuperBlock™ Buffer