missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Chromogenic Endotoxin Quant Kit

A highly sensitive endpoint assay that accurately measures and detects endotoxin (lipopolysaccharide) in a protein, peptide, nucleic acids or antibody sample using the Amebocyte Lysate Assay.

Brand:  Thermo Scientific™ A39552S

Product Code. 15928130

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Highly sensitive Endotoxing detection assay with a broad range — detect as little as 0.01 EU/mL to 1 EU/mL
  • Specific assay — no interference from ß-Glucans, and suitable for wide range of samples, including proteins, vaccine, plasmid, DNA, RNA samples. 
  • Fast — perform assay in as little as 20 minutes
  • End-point chromogenic assay — measure with a standard spectrophotometer or plate reader at 405-410 nm.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Lyophilized amebocyte lysate
• E.coli endotoxin standard
• Chromgenic substrate
• Endotoxin-free water

Store at 2–8°C upon receipt.

0.01 EU/mL
Colorimetric
Pierce™
Endotoxin Quantitation Kit
Kit
0.01 to 0.1 EU/mL, 0.1 to 1.0 EU/mL
End-point Chromogenic
Wet Ice
30 Reactions
30 Microplate-Well Assays Containing 0.01 to 1 Endotoxin Units/mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.