Learn More
Pidotimod, TRC
Product Code. 30009883
Change view
Click to view available options
Quantity:
50 mg
1 g
Unit Size:
1g
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009883 1 g 1g
30006961 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30009883 Supplier Toronto Research Chemicals Supplier No. TRCP4376501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 121808-62-6
Synonym (4R)-3-[(2S)-5-oxopyrrolidine-2-carbonyl]-1,3-thiazolidine-4-carboxylic acid
Specifications
Format Neat
Molecular Weight (g/mol) 244.27
Quantity 1 g
Shipping Condition Room Temperature