Learn More
Eurofins DiscoverX PI3 Kinase (p110δ/p85α) Human Protein
Description
Phosphoinositide 3-kinases (PI3Ks) phosphorylate the 3-prime OH position of the inositol ring of inositol lipids. See MIM 602838. The class I PI3Ks display a broad phosphoinositide lipid substrate specificity and include p110-alpha (MIM 171834), p110-beta (MIM 602925), and p110-gamma (MIM 601232). p110-alpha and p110-beta interact with SH2/SH3-domain-containing p85 adaptor proteins (see MIM 171833) and with GTP-bound Ras.
GenBank NM_005026. The recombinant protein contains the amino acid substitution N253S with respect to this accession number. This conflict is reported inGenBank Y10055 and U57843. The residue coordinates in the native sequence are given / GenBank XM_043865
Spécification
Spécification
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 121.6 kDa (P110δ) and 83.7 kDa (p85α) |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His / Untagged |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | Combined ≥70% |
| Protein | PI3 Kinase (p110δ/p85α) |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.