missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI-103 Hydrochloride, MedChemExpress
Description
PI-103 Hydrochloride is a dual PI3K and mTOR inhibitor with IC50s of 8 nM, 88 nM, 48 nM, 150 nM, 20 nM, and 83 nM for p110α, p110β, p110δ, p110γ, mTORC1, and mTORC2. PI-103 Hydrochloride also inhibits DNA-PK with an IC50 of 2 nM. PI-103 Hydrochloride induces autophagy.
Specifications
Specifications
| Chemical Name or Material | PI-103 Hydrochloride |
| CAS | 371935-79-4 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C19H17ClN4O3 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 4.1 mg/mL (10.65 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | OC1=CC(C2=NC3=C(OC4=C3C=CC=N4)C(N5CCOCC5)=N2)=CC=C1.[H]Cl |
| Molecular Weight (g/mol) | 384.82 |
| Formula Weight | 384.82 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?