Learn More
uvex Phynomic AirLite B ESD Safety Glove
Shop All Uvex Products
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
10 pairs
Beskrivning
- The lightest and most sensitive uvex phynomic cut protection glove in Cut Level B
- Outer material made from innovative Dyneema Diamond 2.0 cut protection fibers, polyamide, elastane and carbon
- Breathable 'airLite' aqua-polymer coating on palm and fingertips
- Safety glove free from glass and steel fibers
- Safety glove with touchscreen function – for use on almost all screens, tablets and mobile phones
- Safety glove with ESD function in line with DIN EN 16350:2014 – protects workpiece against electrostatic charge
- Good cut protection (Level B)
- Good grip in dry and slightly damp areas
- Free from accelerators, and health protection and skin compatibility dermatologically approved (proDERM)
- Very lightweight and breathable safety glove
- Outstanding sensitivity thanks to new breathable 'airLite' aqua-polymer coating combined with fine liner (18 gauge)
- Safety gloves with knitted cuff and ergonomic fit
Specifikationer
Specifikationer
| Certifications/Compliance | EN 388 (3 X 3 2 B) and DIN EN 16350:2014 |
| Coating Material | Aqua-Polymer Foam |
| Color | Light Blue, Black |
| Disposable | Reusable |
| Exterior Finish | Smooth |
| Hand | Ambidextrous |
| Laboratory/Cleanroom Use | Not tested |
| Latex Free | Yes |
| Material | Polyamide, Elastane, Carbon Fiber |
| Powdered | Powder Free |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering