Learn More
Phosphoserine Antibody (PSR-45), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-405
This item is not returnable.
View return policy
Description
Phosphoserine Monoclonal specifically detects Phosphoserine in Human, Rat, Rabbit samples. It is validated for Western Blot, ELISA.
Specifications
| Phosphoserine | |
| Monoclonal | |
| Unconjugated | |
| Phosphoserine; Phosphothreonine | |
| Phosphoserine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| 0.1 mL | |
| Primary | |
| Human, Rat, Rabbit | |
| Ascites |
| Western Blot, ELISA | |
| PSR-45 | |
| Western Blot 1:500-1:1000, ELISA 1:4000 | |
| Mouse | |
| Unpurified | |
| RUO | |
| By ELISA and dot blot, the antibody reacts specifically with phosphorylated serine, both as free amino acid or conjugated to carriers such as BSA or KLH. No cross-reactivity is observed with non-phosphorylated serine, phosphothreonine, phosphotyrosine, AMP or ATP. This antibody has been used in immunoblotting for the localization of some phosphoserine-containing proteins. Certain proteins known to contain phosphorylated serine may not be recognized by this antibody due to steric hindrance of the recognition site. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction