missing translation for 'onlineSavingsMsg'
Learn More

Phosphoenolpyruvic acid tricyclohexylammonium salt

Product Code. 15410227
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410227 25 g 25g
15464419 1 g 1g
15474419 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15410227 Supplier Thermo Scientific Alfa Aesar Supplier No. J61943.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Metabolic intermediate in many kinase reactions | CAS: 35556-70-8 | C21H44N3O6P | 465.572 g/mol

Metabolic intermediate in many kinase reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35556-70-8
Molecular Formula C21H44N3O6P
Molecular Weight (g/mol) 465.57
MDL Number MFCD00004258
InChI Key MJKYGUXBFYGLLM-UHFFFAOYSA-N
Synonym phosphoenolpyruvic acid tris cyclohexylammonium salt, tris cyclohexylamine ; phosphoenolpyruvic acid, phosphoenolpyruvic acid, tris cyclohexylammonium salt, pep-3cha, phosphoenol pyruvate tri cyclohexylammonium salt, phosphoenolpyruvic acid tricyclohexylammonium salt, phosphoenolpyruvic acid tris cyclohexylamine salt, 2-phosphonooxy-2-propenoic acid tri cyclohexylammonium salt, axi c 1/4 +/-ui feminineea ey>>. 1/4 masculine degrees .ni, phospho enol pyruvic acid tri cyclohexylammonium salt enzymatic
PubChem CID 11754241
IUPAC Name 2-(phosphonooxy)prop-2-enoic acid; tris(cyclohexanamine)
SMILES NC1CCCCC1.NC1CCCCC1.NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O

Specifications

Color White
Physical Form Powder
Quantity 25 g
Beilstein 3752336
Formula Weight 465.58
Percent Purity ≥99%
Chemical Name or Material Phosphoenolpyruvic acid tricyclohexylammonium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.