missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-(Ser/Thr) ATM + ATR Substrate Monoclonal Antibody (G.770.10)

Product Code. 11382062
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382062 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11382062 Supplier Invitrogen™ Supplier No. MA514872

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with corresponding nonphosphorylated sequences or with other phospho-Ser/Thr-containing motifs. This antibody preferentially binds peptides and proteins that contain phospho-Ser/Thr preceded by Leu or similar hydrophobic amino acids at the -1 position and followed by Gln at the +1 position.

Ataxia telangiectasia mutated kinase (ATM) and ataxia telangiectasia and Rad3-related kinase (ATR) are related kinases that regulate cell cycle checkpoints and DNA repair. The identified substrates for ATM are p53, p95/NBS1, MDM2, Chk2, BRCA1, CtIP, 4E-BP1, and Chk1. The essential requirement for the substrates of ATM/ATR is S*/T*Q. Hydrophobic amino acids at positions -3 and -1, and negatively charged amino acids at position +1 are positive determinants for substrate recognition by these kinases. Positively charged residues surrounding the S*/T*Q are negative determinants for substrate phosphorylation. The complex phenotype of AT cells suggests that it likely has additional substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-(Ser/Thr) ATM + ATR Substrate
Applications Western Blot
Classification Monoclonal
Clone G.770.10
Concentration 192 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene Alias L(s/t)Q; SQ/TQ
Host Species Rabbit
Immunogen Synthetic phospho-ATM/ATR substrate peptides.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.