missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-PLK1 (Thr210) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16631085
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16631085 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16631085 Supplier Invitrogen™ Supplier No. PA5104633

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of PLK1 only when phosphorylated at Thr210.

PLK1 is a serine/threonine kinase that is thought to function in cell mitosis and regarded as a marker of cell proliferation. It has been found to play a critical role in carcinogenesis. Polo-like kinase proteins acts by binding and phosphorylating proteins are that already phosphorylated on a specific motif recognized by the POLO box domains. Phosphorylates BORA, BUB1B/BUBR1, CCNB1, CDC25C, CEP55, ECT2, ERCC6L, FBXO5/EMI1, FOXM1, KIF20A/MKLP2, CENPU, NEDD1, NINL, NPM1, NUDC, PKMYT1/MYT1, KIZ, PPP1R12A/MYPT1, PRC1, RACGAP1/CYK4, SGOL1, STAG2/SA2, TEX14, TOPORS, p73/TP73, TPT1 and WEE1. Plays a key role in centrosome functions and the assembly of bipolar spindles by phosphorylating KIZ, NEDD1 and NINL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PLK1 (Thr210)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Plk1
Gene Accession No. P53350, Q07832, Q62673
Gene Alias cell cycle regulated protein kinase; Plk; PLK 1; Plk1; PLK-1; plk1.S; plx1; polo (Drosophia)-like kinase; polo like kinase; polo like kinase 1; polo like kinase 1 S homeolog; polo-like kinase; polo-like kinase 1; polo-like kinase homolog; polo-like protein kinase; serine/threonine-protein kinase 13; Serine/threonine-protein kinase PLK1; STPK13; STPK14; Unknown (protein for MGC:133927); XELAEV_18047692mg
Gene Symbols Plk1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PLK1(Accession P53350), corresponding to amino acid residues around phosphorylated Thr210.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18817, 25515, 5347
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.