missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PERK (Thr982) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121311
This item is not returnable.
View return policy
Description
Positive test controls include: 293T. The target is usually found in the following locations: Endoplasmic reticulum membrane, Single-pass type I membrane protein. Immunogen sequence: RHTGQ.
EIF2AK3 (PERK) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK3, signaling is initiated by misfolded proteins in the ER. Defects in EIF2AK3 are the cause of Wolcott-Rallison syndrome (WRS), also known as multiple epiphyseal dysplasia with early-onset diabetes mellitus. WRS is a rare autosomal recessive disorder, characterized by permanent neonatal or early infancy insulin-dependent diabetes and, at a later age, epiphyseal dysplasia, osteoporosis, growth retardation and other multisystem manifestations, such as hepatic and renal dysfunctions, mental retardation and cardiovascular abnormalities.
Specifications
| Phospho-PERK (Thr982) | |
| Polyclonal | |
| Unconjugated | |
| EIF2AK3 | |
| 2.7.11.1; AI427929; EIF2AK3; eukaryotic translation initiation factor 2 alpha kinase 3; eukaryotic translation initiation factor 2-alpha kinase 3; HsPEK; OTTHUMP00000207187; OTTHUMP00000207188; Pancreatic eIF2-alpha kinase; PEK; Perk; PRKR-like endoplasmic reticulum kinase; WRS | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13666, 29702, 9451 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.78 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NZJ5, Q9Z1Z1, Q9Z2B5 | |
| EIF2AK3 | |
| A synthetic phosphorylated peptide around T982 of human PERK (NP_004827.4). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction