missing translation for 'onlineSavingsMsg'
Learn More

Phospho-NIPA (Ser354) Polyclonal Antibody, Invitrogen™

Product Code. 15595691
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15595691 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15595691 Supplier Invitrogen Supplier No. PA538782

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Essential component of an SCF-type E3 ligase complex, SCF(NIPA), a complex that controls mitotic entry by mediating ubiquitination and subsequent degradation of cyclin B1 (CCNB1). Its cell-cycle-dependent phosphorylation regulates the assembly of the SCF(NIPA) complex, restricting CCNB1 ubiquitination activity to interphase. Its inactivation results in nuclear accumulation of CCNB1 in interphase and premature mitotic entry. May have an antiapoptotic role in NPM-ALK-mediated signaling events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-NIPA (Ser354)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZC3HC1
Gene Accession No. Q86WB0, Q80YV2
Gene Alias 1110054L24Rik, NIPA, hNIPA, hematopoietic stem/progenitor cell protein 216, nuclear interacting partner of anaplastic lymphoma kinase (ALK), nuclear interaction partner of ALK, nuclear-interacting partner of ALK, nuclear-interacting partner of anaplastic lymphoma kinase, zinc finge
Gene Symbols ZC3HC1
Host Species Rabbit
Immunogen A synthetic phosphopeptide derived from human NIPA around the phosphorylation site of Ser354 (T-R-SP-W-D)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232679, 296957, 51530
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.