missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-JNK1/JNK2/JNK3 (Thr183, Tyr185) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15574041
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15574041 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15574041 Supplier Invitrogen™ Supplier No. PA536753

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 46 and 54 kDa. Purity is >95% by SDS-PAGE.

C-Jun N-terminal Kinase (JNK) is also known as Mitogen-activated protein kinase (MAPK8). It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predominant forms are JNK1 and JNK2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-JNK1/JNK2/JNK3 (Thr183, Tyr185)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MAPK9
Gene Accession No. P45983, P45984, P49185, P49186, P49187, P53779, Q61831, Q91Y86, Q9WTU6
Gene Alias AI849689; AI851083; C230008H04Rik; c-Jun kinase 2; c-Jun N-terminal kinase 1; c-Jun N-terminal kinase 2; c-Jun N-terminal kinase 3; FLJ12099; FLJ33785; hypothetical protein LOC569698; janus kinase 1; janus kinase 2; JNK; JNK 46; JNK 55; JNK/SAPK alpha; JNK1; JNK1 beta1 protein kinase; JNK1A2; Jnk2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; Jnk3; JNK3 alpha protein kinase; JNK3 beta1 protein kinase; JNK3 beta2 protein kinase; JNK3A; JNK3B1; JNK3B2; JNK-46; JNK-55; Jun kinase; JUN N-terminal kinase; MAP kinase 10; MAP kinase 8; MAP kinase 9; MAP kinase p49 3F12; MAPK 10; MAPK 8; MAPK 9; Mapk10; Mapk8; Mapk9; MGC137545 protein; MGC50974; Mi; mitogen activated protein kinase 10; mitogen activated protein kinase 8; mitogen activated protein kinase 9; mitogen-activated protein kinase 10; Mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; mitogen-activated protein kinase 9; MK10; p493F12; p54 gamma; p54a; p54-alpha; p54aSAPK; p54-beta; p54bSAPK; Phospho-JNK; PRKM10; Prkm8; PRKM9; protein kinase mitogen-activated 8; protein kinase, mitogen-activated 9; SAPb; SAPK; SAPK gamma; SAPK(beta); SAPK/Erk/kinase 2; SAPK1; SAPK1a; SAPK1B; SAPK1c; SAPK-alpha; SAPK-beta; SAPKC; Serk2; si:ch211-247j9.2; stress activated protein kinase alpha II; Stress activated protein kinase beta; stress-activated protein kinase 1; stress-activated protein kinase 1a; Stress-activated protein kinase 1b; stress-activated protein kinase 1c; Stress-activated protein kinase JNK1; stress-activated protein kinase JNK2; Stress-activated protein kinase JNK3; zgc:123234
Gene Symbols MAPK10, MAPK8, MAPK9
Host Species Rabbit
Immunogen Synthetic phosphopeptide derived from human JNK1 around the phosphorylation site of Threonine 183 and Tyrosine 185.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116554, 25272, 26414, 26419, 26420, 50658, 5599, 5601, 5602
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.