missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-IP3 Receptor 1 (Ser1764) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15576981
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15576981 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15576981 Supplier Invitrogen™ Supplier No. PA540296

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with human and mouse based on sequence homology.

Intracellular channel that mediates calcium release from the endoplasmic reticulum following stimulation by inositol 1,4,5-trisphosphate. Involved in the regulation of epithelial secretion of electrolytes and fluid through the interaction with AHCYL1. Plays a role in ER stress-induced apoptosis. Cytoplasmic calcium released from the ER triggers apoptosis by the activation of CaM kinase II, eventually leading to the activation of downstream apoptosis pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-IP3 Receptor 1 (Ser1764)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4
Gene Itpr1
Gene Accession No. P11881, P29994, Q14643
Gene Alias ACV; CLA4; D6Pas2; ENSMUSG00000072853; Gm10429; I145TR; inositol 1,4,5-triphosphate receptor 1; inositol 1,4,5-triphosphate receptor, type 1; inositol 1,4,5-trisphosphate receptor 1; inositol 1,4,5-trisphosphate receptor type 1; inositol 1,4,5-trisphosphate receptor, type 1; inositol 1,4,5-trisphosphate-binding protein P400; InsP>3>R type I; Insp3r; InsP3R type I; insP3R1; IP3 receptor; IP3 receptor isoform 1; Ip3r; IP-3-R; IP3R 1; IP3R1; Itpr1; Itpr-1; opisthotonus; opt; P400; Pcd6; Pcp1; Pcp-1; PPP1R94; protein PCD-6; protein phosphatase 1, regulatory subunit 94; Purkinje cell protein 1; SCA15; SCA16; SCA29; SI-SIII-SIIA; Type 1 inositol 1,4,5-trisphosphate receptor; type 1 InsP3 receptor
Gene Symbols Itpr1
Host Species Rabbit
Immunogen Synthesized peptide derived from human IP3R-I around the phosphorylation site of S1764.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16438, 25262, 3708
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.