missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Ezrin (Thr567) Polyclonal Antibody

Description
Antibody detects endogenous levels of Ezrin only when phosphorylated at Threonine 567.
The cytoplasmic peripheral membrane protein encoded by this gene functions as a protein-tyrosine kinase substrate in microvilli. As a member of the ERM protein family, this protein serves as an intermediate between the plasma membrane and the actin cytoskeleton. This protein plays a key role in cell surface structure adhesion, migration and organization, and it has been implicated in various human cancers. A pseudogene located on chromosome 3 has been identified for this gene. Alternatively spliced variants have also been described for this gene.
Specifications
Specifications
| Antigen | Phospho-Ezrin (Thr567) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Ezr |
| Gene Accession No. | P15311, P26040, P31977 |
| Gene Alias | AW146364; CVIL; CVL; Cytovillin; cytovillin 2; DKFZp762H157; epididymis secretory protein Li 105; ezr; ezr.L; ezr.S; EZRI; Ezrin; ezrin L homeolog; ezrin L homeolog; ezrin S homeolog; FLJ26216; HEL-S-105; MGC1584; p81; R75297; VIL2; villin 2; villin 2 (ezrin); villin-2; XELAEV_18027043mg |
| Gene Symbols | Ezr |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?