missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599187
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CDK7.
The trimeric complex of CDK7, cyclin H and MNAT1, functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This complex is thought to serve as a direct link between the regulation of transcription and the cell cycle. The bound complex is important in the normal double-strand break repair function of breast cancer, type 1 (BRCA1). CDK7 may be a target of germline cancer-inducing mutations.
Specifications
| CDK7 | |
| Polyclonal | |
| Unconjugated | |
| CDK7 | |
| 39 KDa protein kinase; 39 protein kinase; AI323415; AI528512; C230069N13; CAK; CAK1; CDK; CDK7; CDK-activating kinase; CDK-activating kinase 1; Cdkn7; cell division protein kinase 7; CR4 protein kinase; CRK4; CRK4 PK (CDC2-related-kinase-4 protein kinase); cyclin dependent kinase 7; cyclin-dependent kinase 7; cyclin-dependent kinase 7 (homolog of Xenopus MO15 cdk-activating kinase); cyclin-dependent kinase 7 (MO15 homolog Xenopus laevis cdk-activating kinase); cyclin-dependent kinase 7 (MO15 homolog, Xenopus laevis, cdk-activating kinase); EC 2.7.11.22; EC 2.7.11.23; HCAK; homolog of Xenopus MO15 Cdk-activating kinase; kinase subunit of CAK; MO15; Mpk-7; OTTHUMP00000222032; OTTHUMP00000222033; p39 Mo15; p39MO15; Protein-tyrosine kinase MPK-7; serine/threonine kinase stk1; serine/threonine protein kinase 1; serine/threonine protein kinase MO15; Serine/threonine-protein kinase 1; STK1; TFIIH basal transcription factor complex kinase subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1022, 12572 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P50613, Q03147 | |
| CDK7 | |
| A synthesized peptide derived from human CDK7(Accession P50613), corresponding to amino acid residues K139-M189. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction