missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Acetyl-CoA Carboxylase (Ser80) Polyclonal Antibody, Invitrogen™

Product Code. 15505011
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15505011 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15505011 Supplier Invitrogen Supplier No. PA538079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Catalyzes the rate-limiting reaction in the biogenesis of long-chain fatty acids. Carries out three functions: biotin carboxyl carrier protein, biotin carboxylase and carboxyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Acetyl-CoA Carboxylase (Ser80)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACACA
Gene Accession No. Q13085, Q5SWU9, P11497
Gene Alias ACAC, ACACA, ACC-alpha, ACCA, Acetyl-CoA carboxylase 1, EC 6.3.4.14, EC 6.4.1.2
Gene Symbols ACACA
Host Species Rabbit
Immunogen A synthetic phosphopeptide derived from human acetyl-CoA carboxylase around the phosphorylation site of Ser80 (S-M-SP-G-L)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107476, 31, 60581
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.