Learn More
Palintest™ Phosphate HR Test Kit
Product Code. 11973647
Click to view available options
Range:
0 to 100mg/L
Unit Size:
Pack of 50
Product Code. 11973647
Brand: Palintest™ PM114
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
No. of Tests 50
Quantity 50 Pack
Range 0 to 100mg/L
Sufficient For 50 Tests
Type Phosphate HR Test Kit
compliance-icons compliance-icons
Product Identifier
  • photometric test kit phosphate 0-100PPM
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
Hazard Statement
  • H318-Causes serious eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : sodium metabisulphite, boric acid, sodium hydrogen sulphate,ammonium molybdate,ammonium metavanada