Learn More
Phosphate buffer solution, pH 7.2, Honeywell Fluka™
Product Code. 15633550 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15633550 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15633550 Supplier Honeywell-Fluka Supplier No. 319252500ML
Request Bulk or Custom Format
Specifications
Chemical Name or Material Phosphate Buffer Solution Ph 7.2
Density 0.995g/mL at 25°C
Quantity 500 mL
Packaging Poly Bottle
UN Number NONH for all modes of transport