missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHKG2 Monoclonal Antibody (OTI2E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800343S
This item is not returnable.
View return policy
Description
PHKG2 Monoclonal antibody specifically detects PHKG2 in Human samples. It is validated for Western BlotSpecifications
| PHKG2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P00488 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2162 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI2E5 | |
| Unconjugated | |
| F13A1 | |
| 1500017I02Rik; GSD9C; MGC129049 protein; phkg2; PHK-gamma-LT; PHK-gamma-T; Phosphorylase b kinase gamma catalytic chain, liver/testis isoform; phosphorylase b kinase gamma catalytic chain, testis/liver isoform; phosphorylase kinase; phosphorylase kinase catalytic subunit gamma 2; phosphorylase kinase gamma subunit 2; phosphorylase kinase subunit gamma 2; phosphorylase kinase subunit gamma-2; phosphorylase kinase, gamma 2 (testis); Phosphorylase kinase, gamma 2 (testis/liver); PSK-C3; serine/threonine-protein kinase PHKG2; similar to phosphorylase kinase, gamma 2 (testis); zgc:55863; zgc:55863 protein | |
| Full length human recombinant protein of human F13A1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction