missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHKG2 Monoclonal Antibody (OTI2E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800343
This item is not returnable.
View return policy
Description
Phosphorylase b kinase catalyzes the phosphorylation of serine in certain substrates, including troponin I. This isozyme may regulate glycogeneolysis in the testis.Specifications
| PHKG2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P15735 | |
| PHKG2 | |
| Human recombit protein fragment corresponding to amino acids 1-216 of human PHKG2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI2E5 | |
| Unconjugated | |
| PHKG2 | |
| GSD9C | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5261 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction