missing translation for 'onlineSavingsMsg'
Learn More

Phenylglyoxylic acid, 98%

Product Code. p-7043407
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11390144

Brand: Thermo Scientific Alfa Aesar L00648.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-73-4 | C8H6O3 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-73-4
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00002575
InChI Key FAQJJMHZNSSFSM-UHFFFAOYSA-N
Synonym benzoylformic acid, phenylglyoxylic acid, oxophenylacetic acid, benzeneglyoxylic acid, phenylgloxylic acid, 2-phenylethanoic acid, oxo phenyl acetic acid, benzoylformate, phenylglyoxalate, phenyloxoacetic acid
PubChem CID 11915
ChEBI CHEBI:18280
IUPAC Name 2-oxo-2-phenylacetic acid
SMILES OC(=O)C(=O)C1=CC=CC=C1

Specifications

Melting Point 62°C to 67°C
Density 1.38
Boiling Point 147°C to 151°C (12 mmHg)
Quantity 5 g
Beilstein 606718
Formula Weight 150.13
Percent Purity 98%
Chemical Name or Material Phenylglyoxylic acid
compliance-icons
Product Identifier
  • Phenylglyoxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Benzoylformic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.