missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetaldehyde dimethyl acetal, 98%, Thermo Scientific™

Product Code. 10306970 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10306970

Brand: Thermo Scientific Acros 130271000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 101-48-4
Molecular Weight (g/mol) 166.22
MDL Number MFCD00008487
InChI Key WNJSKZBEWNVKGU-UHFFFAOYSA-N
Synonym 2,2-dimethoxyethyl benzene, phenylacetaldehyde dimethyl acetal, viridine, hyscylene p, 1,1-dimethoxy-2-phenylethane, benzene, 2,2-dimethoxyethyl, phenacetaldehyde dimethyl acetal, padma, 2,2-dimethoxy-1-phenylethane, phenylacetic aldehyde dimethyl acetal
PubChem CID 60995
IUPAC Name 2,2-dimethoxyethylbenzene
SMILES COC(CC1=CC=CC=C1)OC

Specifications

Acidity 1mg KOH/g max.
Color Colorless to Yellow
Density 1.0040g/mL
Boiling Point 219°C to 221°C (754.0 mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H14O2
Linear Formula C6H5CH2CH(OCH3)2
Refractive Index 1.4930 to 1.495
Quantity 100 g
Beilstein 07, 293
Specific Gravity 1.004
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in 2 parts of 70% alcohol
Formula Weight 166.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Phenylacetaldehyde dimethyl acetal, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.