missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Phenols Cuvette Tests

Product Code. 11311443
Click to view available options
No. of Tests:
24
Unit Size:
24 tests
This item is not returnable. View return policy

Product Code. 11311443

Brand: Hach Lange™ LCK345

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

No. of Tests 24
Range 0.05-5mg/L
Storage Requirements 15°C to 25°C
Test Method 4-Nitroaniline
Type Phenols Cuvette Test
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 24 Tests Phenols cuvette test measuring range 0.05-5.0 mg/l
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid, sodium nitrate, sodium carbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.