Learn More
Phenolphthalein Indicator in Ethanol/Water, Chem Lab™
Product Code. 11923333
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11923333 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11923333 Supplier Chem-Lab Supplier No. CL06.0608.1000
Specifications
Melting Point -114°C
Solubility Information Soluble in water
Odor Alcoholic
Color Colorless
Quantity 1 L
Physical Form Liquid
compliance-icons
Product Identifier
  • Phenolphtalein 5 g/l in ethanol/water (1/1) indicator solution
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: phenolphthalein, ethanol