missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Phenol Reagent Test Kits
Click to view available options
:
Reagent Test; Detection Range: 0.002 to 0.100mg/L C6H5OH, 0.025 to 5mg/L C6H5OH
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifications
Specifications
| Analytes | Indophenol blue (Berthelot reaction) |
| For Use With (Application) | For waste water, surface water |
| For Use With (Equipment) | For PhotoLab™ S12, PhotoLab™ Spectral photometers |
| Includes | Certificates |
| No. of Tests | 50 |
| Range | 0.002 - 0.100mg/L for 20mm cuvette; 0.025 - 5.00mg/L for 10, 20, 50mm cuvette |
| Sufficient For | 50 Tests |
| Test Method | Photometric |
| Type | Ammonium (NH4) cuvette test kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction