missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene-2-silyl Ether, TRC

Product Code. 30412531
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412531 5 mg 5mg
30424687 2.5 mg 2.5mg
30428255 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30412531 Supplier Toronto Research Chemicals Supplier No. TRCP2948305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenanthrene-2-silyl Ether
Analyte or Component Names Phenanthrene-2-silyl Ether
Shipping Condition Room Temperature
Molecular Formula C20 H24 O Si
Synonym t-Butyldimethyl(phenanthren-2-yloxy)silane
InChI Formula InChI=1S/C20H24OSi/c1-20(2,3)22(4,5)21-17-12-13-19-16(14-17)11-10-15-8-6-7-9-18(15)19/h6-14H,1-5H3
Solution Type Neat
SMILES CC(C)(C)[Si](C)(C)Oc1ccc2c(ccc3ccccc23)c1
IUPAC Name tert-butyl-dimethyl-phenanthren-2-yloxysilane
Industry Type Pharmaceutical
Molecular Weight (g/mol) 308.49
Quantity 5 mg
Formula Weight 308.16
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.