Learn More
Motic™ Phase Stop 0.5 Ph0 (4X)
Product Code. 15445362
Click to view available options
For Use With (Equipment):
AE31E Microscope, 4X Phase Objective
Unit Size:
Each
Product Code. 15445362
Brand: Motic™ 1101000202411
Only null left
Add to basket
Add to basket
Use phase contrast to improve visualization of transparent specimens. Motic™ Phase Stop 0.5 Ph0 (4X) is an annular light stop that fits in your condenser.
  • Boost contrast and detail discrimination without staining or fixing, thus permitting live cell study.
  • Must be matched and aligned to the objective phase plate.
Specifications
Type Phase Stop Annulus
Description 0.5 Ph0 (4X)
For Use With (Equipment) AE31E Microscope, 4X Phase Objective