Learn More
Motic™ Phase Stop 0.5 Ph0 (4X)
Click to view available options
For Use With (Equipment):
AE31E Microscope, 4X Phase Objective
Unit Size:
Each
Description
- Boost contrast and detail discrimination without staining or fixing, thus permitting live cell study.
- Must be matched and aligned to the objective phase plate.
Specifications
Specifications
| Type | Phase Stop Annulus |
| Description | 0.5 Ph0 (4X) |
| For Use With (Equipment) | AE31E Microscope, 4X Phase Objective |