Learn More
Motic™ Phase Centering Telescope
Description
- Use to align annulus and objective for proper phase contrast viewing.
- Improve visualization of transparent specimens.
Specifications
Specifications
| For Use With (Equipment) | DMW-BI, B3, B1, B1-223ASC, B3-223PL Microscopes |
| Diameter (Metric) | 23.2 mm |